Mesh Overscreen for Sanitary Strainer 3" dia; 100×100 Mesh
Supplier: RUBBERFAB GASKET & MOLDING
VWR Item#MFLX29590-37
Your Organization's Price:
Retrieving
Protect products and expensive process equipment from coarse or fine particulates.
  • Mesh overscreens are available in a variety of mesh sizes to meet your specific application
  • Overscreens are hemmed smooth with no stray wire ends
  • Overscreens fit both in-line and side-inlet sanitary strainer assemblies
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.