ODE Solenoid Valve NSF-Certified 1/8" BSP(F) NC; PPS body; KFM Seal/PPS Seat
Supplier: FLUID CONVEYING PRODUCTS
VWR Item#MFLX08617-94
Your Organization's Price:
Retrieving
Wetted materials fully compliant with NSF standards to ensure water and food safety.
  • Valves create zero differential pressure
  • Integrated mounting holes for ease of installation or replacement
  • Low actuation wattage required, resulting in low energy consumption and operating cost
These two-way normally closed solenoid valves are used in sterilization, process, and vending of liquids, beverages, and food where NSF certification is required. High-quality virgin materials provide extended service life, and high pressure and temperature resistance The orifice food-grade sealing materials ensure compatibility with media and sealing for precise flow management.