Spirax Sarco Cast Iron "Y" Strainer, 2" NPT(F); 1/Each
Supplier: Avantor
Keep steam space free of condensate to provide maximum system efficiency.
  • Constructed of durable cast iron
Protect your pipeline from debris with these strainers.