Spirax Sarco Cast iron inverted bucket steam trap; 3/4" NPT(F), 75 psig
Supplier: Avantor
Keep steam space free of condensate to provide maximum system efficiency.
- All stainless steel working parts provide good resistance to corrosion and wear
- Inverted bucket steam traps rely on the difference in density between steam and condensate to drain condensate as it forms.
- Condensate is discharged close to steam temperature
<B>Typical applications:</B><br />Steam main drip stations<br />Laundry equipment<br />Industrial dryers<br />Storage tanks