Masterflex® L/S® Easy-Load® Head with Open-Head Sensor for High-Performance Precision Tubing, PARA Housing, SS Rotor
Supplier: Avantor Fluid Handling
Save time with fast tubing changes and reduced maintenance.
- New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
- Rotor is constructed of stainless steel (SS) for enhanced durability
- Pump heads are stackable for multichannel applications
- Compact design for efficient use of space
- Suitable for use with L/S® High-Performance Precision pump tubing sizes
- Models with open-head interlock sensor stop drive motor when pump head is open (when used with sensor-equipped drives)
This Easy-Load® pump head accepts high-performance precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 15 or L/S 24.