Flowline DS14-00 EchoPod (Red) Switch & Cont.-1.25m range, 1" NPT
Supplier: FLOWLINE
An integrated control automation solution that provides noncontact level measurement with switch and control capabilities.
- Compact design allows for installation in applications with limited mounting options and space
- Measures dirty, sticky and corrosive media without affecting product performance
- Advanced Digital Signal Processing provides surface imaging in applications with obstacles in the beam width
- A single unit can provide switch, control and measurement capabilities simultaneously or individually
EchoPod's relays can be configured on a single set point alarm or latched on two sets for automatic fill or empty in simplex or duplex control modes with fail-safe logic. The embedded controller can replace external control hardware. The sensor is well suited for a wide range of corrosive, sticky or dirty type media. EchoPod is broadly selected for day tanks, sumps, IBC totes, lift stations and mini-bulk storage applications.<br /><br /><B>DL24 Ultrasonic Multifunction Transmitter, Switch & Controller</B><br />The general purpose, ultrasonic sensor provides non-contact level detection up to 98.4'' (2.5m) with 4 SPST 60 VA 1A relays and a two-wire 4-20 mA level measurement output.<br /><br /><B>DL14 Ultrasonic Multifunction Transmitter, Switch & Controller</B><br />The general purpose, ultrasonic sensor provides non-contact level detection up to 49.2'' (1.25m) with 4 SPST 60 VA 1A relays and a two-wire 4-20 mA level measurement output.<br /><br /><B>DS14 Ultrasonic Switch & Controller with (4) Relays</B><br />The general purpose, ultrasonic sensor provides non-contact level detection up to 49.2'' (1.25m) with 4 SPST 60 VA 1A relays. The DS14 directly replaces contact switches connected to a PLC with 1-4 discrete points for easy design conversion or field retrofit.<br /><br /><B>DX10 Ultrasonic Sensor with Voltage & Frequency Output</B><br />The general purpose, DX10 ultrasonic sensor provides non-contact level measurement up to 49.2'' (1.25m) with a 0-5 VDC, 0-10 VDC or 976-2000 Hz. frequency output. Select the voltage output to directly replace a contact measurement sensor connected to a PLC analog input. Select the frequency output to replace a contact measurement sensor or switch assembly, and eliminate the cost of an analog input card by connecting the DX10 to a PLC discrete input.<br /><br /><B>DL10 Ultrasonic Transmitter with 4-20 mA Output</B><br />The general purpose, DL10 ultrasonic sensor provides non-contact level measurement up to 49.2'' (1.25m) with a 4-20 mA output. The DL10 directly replaces contact measurement sensors connected to a PLC analog input card for easy design conversion or field retrofit. The two-wire, 4-20 mA output is ideal for longer signal distances up to 1000'.
Note: WebCal software is required for configuration and setup of EchoPod. Download WebCal for free.