Masterflex® PTFE-Diaphragm Pump Head, 10 to 100 mL/min
Supplier: Avantor Fluid Handling
Compatible with most Masterflex® L/S® drives.
- PTFE and glass wetted parts help maintain fluid purity
- Ideal for pumping through filters or into pressurized systems
- Check valve on inlet side of pump holds prime for up to two hours when pump is off
Note: Do not use these pump heads with Masterflex® drives requiring a long-shaft pump head.
<b>Delivery information: </b>Includes mounting tools, coupling hardware, and service kit.