Hand-Operated Lever drum pump, 4 strokes per gallon, steel pump body
Supplier: Action Pump
VWR Item#MFLX06511-00
Your Organization's Price:
Retrieving
Metal-body pumps for use in tight work areas where drums containing flammables are stored.
  • Curved discharge spout for convenient hose extension
  • Designed for use in industrial, agricultural, and automotive industries
  • Model 06511-00 is ideal for use with high-viscosity fluids
  • Materials vary from Stainless steel to steel bodies with PTFE or Delrin acetal construction allow for easy handling of the toughest fluids
Lever drum pump model 06511-05 is able to dispense kerosene, antifreeze, lightweight oils, non-water-based and non-corrosive detergents, waxes, or disinfectants. High viscosity model 06511-00 can be used with paraffin, motor oil, cutting oil and other viscous lubricants. This pump comes with a high-angle discharge spout with screen and a curved discharge spout.
Steel pump body, Delrin acetal piston, neoprene seals