Masterflex® L/S® High-Performance Precision Pump Tubing, PharMed® BPT, L/S 35; 25 ft
Supplier: VWR International
Protect your light-sensitive samples.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Single-use custom lengths and configurations available to minimize downtime
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Tubing Size: 35
<B>Applications</B>: Cell and tissue culture work, pharmaceutical, light-sensitive samples<br /><br /><B>Characteristics</B>: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
See 96122-24 for the irradiated version of this tubing.