Masterflex® Ismatec® Pump Tubing, Tygon® S3™ E-LFL, 1.42 mm ID; 100 ft
Supplier: VWR International
Longest life of all Tygon® formulations.
- Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Tubing Inner Diameter: 1.42 mm
<B>Applications</B>: General laboratory use<br /><B>Characteristics</B>: Long life, won’t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, transparent