Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® S3™ E-LFL , L/S 36; 25 ft
Supplier: VWR International
VWR Item#MFLX06440-36
Your Organization's Price:
Retrieving
Longest life of all Tygon® formulations.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 36
<B>Applications</B>: General laboratory use<br /><B>Characteristics</B>: Long life, won’t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, transparent