Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex® ULTRA, L/S 15; 25 ft
Supplier: VWR International
VWR Item#MFLX06434-15
Your Organization's Price:
Retrieving
Longest life of any C-Flex® formulation.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 15
<B>Applications</B>: General-purpose lab uses requiring high performance and long life<br /><B>Characteristics</B>: Exceptional tensile and tear strength, good biocompatibility, low spallation, reduced residue and low gas permeability, smooth surface, translucent