PVC Stopcock w/ Flow Indication, 1/4-1/2" ID×1/4-1/2" ID
Supplier: ASAHI/AMERICA INC
Easily control flow with flow indicator handle.
- Constructed of durable PVC and EPDM
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120 °F (49 °C); maximum vacuum is 29.9" Hg.