Masterflex® Brass Metering Valve, straight flow, 350 mL/min
Supplier: VWR International
Choose from different metal materials based on your application.
- Brass and 316 Stainless Steel material options available
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.