Masterflex® High-Resolution 316 SS Metering Valve, straight flow, 650 mL/min
Supplier: AALBORG INSTRUMENTS & CONTROL
Choose from different metal materials based on your application.
- Brass and 316 Stainless Steel material options available
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.