Masterflex® High-Resolution 316 SS Metering Valve, straight flow, 30 mL/min
Supplier: VWR International
VWR Item#MFLX03214-93
Your Organization's Price:
Retrieving
Choose from different metal materials based on your application.
  • Brass and 316 Stainless Steel material options available
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.