Masterflex® High-Resolution Brass Metering Valve, straight flow, 12 mL/min
Supplier: AALBORG INSTRUMENTS & CONTROL
VWR Item#MFLX03214-82
Your Organization's Price:
Retrieving
Choose from different metal materials based on your application.
  • Brass and 316 Stainless Steel material options available
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.