Deck-Mount Swing-Out Eyewashes, WaterSaver Faucet
Supplier: WaterSaver Faucet
CTEW805 CTEW806 EW805 EW805LH EW806 EW806LH
VFCTEW805EA 922.17 CAD
VFCTEW805 VFCTEW806 59380-018 59380-020 59380-022 59380-024
Deck-Mount Swing-Out Eyewashes, WaterSaver Faucet
First Aid Supplies Eyewash Stations and Eyewash Bottles
Featuring the GS-Plus 'Gentle Spray' heads, these eyewashes deliver a soft, wide stream of water to quickly and gently rinse contaminants away.
  • Includes self-adjusting flow controls, impact-resistant polypropylene body, and flip-top dust covers
  • Auto-flow models are activated when the head assembly is swung from storage to operation position
  • An ANSI-compliant eyewash sign is included with all units
Flag-handle models are activated when the head assembly is swung from storage to operation position and the flag handle is pushed. LDPE filter removes scale,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable