VWR® Modular Workstations with Upper Support, W×D×H 60×30×36"
Supplier: VWR International
VF-U603036E VF-U603036EB VF-U603036L VF-U603036PB
VF-U603036EEA 7188.97 CAD
VF-U603036E VF-U603036EB VF-U603036L VF-U603036PB
VWR® Modular Workstations with Upper Support, W×D×H 60×30×36"
Furniture Tables and Workbenches
VWR® modular work benches are designed for a wide range of applications in laboratory and industrial environments including biotech, pharmaceutical, analytical, material handling, distribution, and assembly.
  • Can be custom configured
  • Shipped with assembled frame
  • Overhead storage capability with addition of uprights
Standard work benches handle 1000lbs
The addition of two uprights (unistruts) to the VWR® Basic Work Bench provides overhead storage capability. The upper support bench includes 4 standard leveler bench feet, gussets and light neutralMore Product Information