Safe-T-Zone® Select Series Combination Shower, Model SE-616, Speakman®
Supplier: Speakman
SE-616
VF-SE-616EA 6626.44 CAD
VF-SE-616
Safe-T-Zone® Select Series Combination Shower, Model SE-616, Speakman®
First Aid Supplies Showers
Speakman® Safe-T-Zone® Select Series combination shower and eyewash unit features a combination eyewash with two yellow aerated nylon spray outlets and a rectangular bowl.
  • Showerhead is activated by a high visibility yellow pull rod
The outlets and bowl are combined on a DuraJade™ powder-coated, galvanized steel stanchion with an impeller action deluge showerhead. The Model SE-616 combination shower is supplied with an SE-505 eyewash with stainless steel bowl.
Certifications: Meets ANSI 358.1.