VWR® Lab Tables
Supplier: VWR International
VB362436-GK VB362448-GK VB362460-GK VB362472-GK VB363036-GK VB363048-GK VB363060-GK VB363072-GK VB363636-GK VB363648-GK VB363660-GK VB363672-GK VB36R012436-BK VB36R012448-BK VB36R012460-BK VB36R012472-BK VB36R013036-BK VB36R013048-BK VB36R013060-BK VB36R013072-BK VB36R013636-BK VB36R013648-BK VB36R013660-BK VB36R013672-BK WB363660-GK WB363072-GK WB362460-GK WB363036-GK WB363636-GK WB362436-GK WB363048-GK WB362472-GK WB362448-GK WB363672-GK WB363060-GK WB363648-GK VB36R022436-BK VB36R023660-BK VB36R023672-BK VB36R022448-BK VB36R022460-BK VB36R022472-BK VB36R023036-BK VB36R023048-BK VB36R023060-BK VB36R023072-BK VB36R023636-BK VB36R023648-BK
VB362436-GKEA 3404.3 CAD
VB362436-GK VB362448-GK VB362460-GK VB362472-GK VB363036-GK VB363048-GK VB363060-GK VB363072-GK VB363636-GK VB363648-GK VB363660-GK VB363672-GK VB36R012436-BK VB36R012448-BK VB36R012460-BK VB36R012472-BK VB36R013036-BK VB36R013048-BK VB36R013060-BK VB36R013072-BK VB36R013636-BK VB36R013648-BK VB36R013660-BK VB36R013672-BK WB363660-GK WB363072-GK WB362460-GK WB363036-GK WB363636-GK WB362436-GK WB363048-GK WB362472-GK WB362448-GK WB363672-GK WB363060-GK WB363648-GK VB36R022436-BK VB36R023660-BK VB36R023672-BK VB36R022448-BK VB36R022460-BK VB36R022472-BK VB36R023036-BK VB36R023048-BK VB36R023060-BK VB36R023072-BK VB36R023636-BK VB36R023648-BK
VWR® Lab Tables
Furniture Tables and Workbenches
A four-leg, flexible lab table system designed for any application.
  • Powder coated steel
  • Chemical resistant epoxy resin
  • Optional shelving
  • Optional casters
  • Accepts suspended cabinets
  • Accepts fixtures (with uprights)
  • 1000 lbs. capacity
This table system includes a number of accessories that can be added very easily, creating your ideal workstation.
Certifications: SEFA tested (SEFA 3 and 10).
Ordering information: Work tops and table frames are sold separately. Tables shipped knocked down, and will need assembly. Professional Assembly is highly recommended.
Order Now
SPECIFICATIONS
Environmentally Preferable