CleanTips® Alpha® ESD Long Handle Swab, Texwipe®
Supplier: Texwipe
TX761D
TWTX761DCS 661.55 CAD
TWTX761D
CleanTips® Alpha® ESD Long Handle Swab, Texwipe®
Swabs Swabs for Cleaning
The Alpha® ESD Swab with Long Handle is an excellent dissipative swab for cleaning surfaces and hard-to-reach areas
This swab can replace long-handled cotton swabs in applications requiring superior cleanliness. The cleanroom laundered, knitted, Alpha® polyester head is extremely clean. A long handle and flexible internal head paddle provide good support. Swab handle is made of low-contamination Transplex®, a unique plastic alloy that safely dissipates electric charges without contaminating additives. Swab handle bears the ESD-safe imprint.
Ordering information: Available in bag or case quantities.