CleanTips® Cotton Foam-Covered Swab, Texwipe®
Supplier: Texwipe
TX720B
TWTX720BCS 1163.46 CAD
TWTX720B
CleanTips® Cotton Foam-Covered Swab, Texwipe®
Swabs Swabs for Cleaning
The Foam-Covered Cotton Swab is a highly sorbent, nonabrasive, general-purpose swab for cleaning surfaces and hard-to-reach areas
Also ideal for cleaning with solvents such as IPA, applying lubricants and other liquids, and removing excess materials. Its open-cell polyurethane foam head possesses excellent absorptive capacity and cushioning while an internal cotton bud provides support and additional sorbency. Good chemical resistance. Soft and nonabrasive. The long hardwood handle allows the head to reach recessed areas and ensures good control and ease of use. Low particle and fiberMore Product Information