CleanTips® Cotton Wrapped Mid-Size Swab, Texwipe®
Supplier: Texwipe
TX705
TWTX705CS 414.49 CAD
TWTX705
CleanTips® Cotton Wrapped Mid-Size Swab, Texwipe®
Swabs Swabs for Cleaning
The Wrapped Cotton Swab, made from 100% cotton fiber, is a highly sorbent utility swab for cleaning surfaces and hard-to-reach areas
It is also ideal for a variety of industrial and manufacturing applications, such as the application of solvents and cleaning solutions. Its cotton head possesses excellent sorbency, cushioning, and heat resistance. The long hardwood handle allows the head to reach recessed areas. Highly absorbent swab is low in particulates and withstands strong solvents. This swab also has the advantage of being seamless, no edges
Ordering information: Available in 1,000More Product Information