SureSTART™ High Recovery Glass Screw Top 1.7 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PSV9-V1 6PSV9-V1F 6PSV9-V1T
TS6PSV9-V1PK 324.93 CAD
TS6PSV9-V1 TS6PSV9-V1F TS6PSV9-V1T
SureSTART™ High Recovery Glass Screw Top 1.7 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 1.7 ml high recovery glass screw top microvials, performance Level 3, when you need to maximize the injection volume for <2 ml samples.
With outer dimensions of 12×32 mm, these clear or amber glass microvials have an internal conical reservoir that provides a total usable volume of 1.5 ml and a residual volume of 4 µl. This is a Performance Level 3 vials, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyzes.