SureSTART™ MSCERT+ Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PMCK30LVW 6PMCK341W 6PMCK351W 6PMCK35W 6PMCK36LVW 6PMCK39TR 6PMCK40W 6PMCK41W
TS6PMCK30LVWPK 254.14 CAD
TS6PMCK30LVW TS6PMCK341W TS6PMCK351W TS6PMCK35W TS6PMCK36LVW TS6PMCK39TR TS6PMCK40W TS6PMCK41W
SureSTART™ MSCERT+ Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Vials
Meet your certification and documentation needs with our level 3 Thermo Scientific™ SureSTART™ MSCERT+ screw vial and cap kits. Select level 3 products for ultimate performance and sensitivity for your methods, while maintaining the highest levels of sample security and integrity.
Choose from a variety of kits containing short thread screw vials with marking spots in clear or amber glass, and polypropylene caps with silicone/PTFE septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Select kits with SureStop™ vials and/or AVCS™ screw caps forMore Product Information