SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
6AMCC20-ST3 6AMCC20-ST3T 6AMCC20-STH 6AMCC20-STHF 6ACC20-PFT 6ACC20-PFTT 6ACC20-ST1 6ACC20-ST1F 6ACC20-ST1T 6AMCC20-STHT 6APRCC20-ST1 6APRCC20-ST1F 6APRCC20-ST1T
TS6AMCC20-ST3PK 249.15 CAD
TS6AMCC20-ST3 TS6AMCC20-ST3F TS6AMCC20-ST3T TS6AMCC20-STH TS6AMCC20-STHF TS6ACC20-PFT TS6ACC20-PFTF TS6ACC20-PFTT TS6ACC20-ST1 TS6ACC20-ST1F TS6ACC20-ST1T TS6AMCC20-STHT TS6APRCC20-ST1 TS6APRCC20ST1F TS6APRCC20ST1T
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 20 mm crimp caps with crimp vials that have a 20 mm opening, including our SureSTART 10 ml and 20 ml glass crimp top headspace vials (level 2). Select level 2 products for high-throughput applications, when robustness and reproducibility are key.
Choose from aluminum and magnetic steel caps with silicone/PTFE or butyl/PTFE septa of various hardness values designed to reduce autosampler needle issues. Caps with pressure release technology will split to allow the safe release of high internal vial pressures. Use Thermo Scientific™ manual or electronicMore Product Information