SureSTART™ Specification Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications
Supplier: Thermo Scientific
6AK92W 6AK93BW 6AK94W 6AK95W 6AK592W 6AK594W 6AK599W 6AK78BW 6AK80W 6AK82W 6AK86W 6AK180W 6AK186W 6AK192W 6AK195W 6AK188W 6AK196W
TS6AK92WPK 92.58 CAD
TS6AK92W TS6AK93BW TS6AK94W TS6AK95W TS6AK592W TS6AK594W TS6AK599W TS6AK78BW TS6AK80W TS6AK82W TS6AK86W TS6AK180W TS6AK186W TS6AK192W TS6AK195W TS6AK188W TS6AK196W
SureSTART™ Specification Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications
Vials
Meet your certification and documentation needs with our level 2 Thermo Scientific™ SureSTART™ specification certified screw vial and cap kits. Select level 2 products for high throughput applications, when robustness and reproducibility are key.
  • Pre-slit and bonded septa options are available
Choose from a variety of kits containing short thread screw vials with marking spots in clear or amber glass and polypropylene caps with silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Select kits with SureStop™ vialsMore Product Information