Vinpocetine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

V0115-100MG V0115-1G
TCV0115-100MGEA 135.56 CAD
TCV0115-100MG TCV0115-1G
Vinpocetine ≥98.0% (by HPLC, titration analysis)
Vinpocetine

CAS Number: 42971-09-5
MDL Number: MFCD00211233
Molecular Formula: C22H26N2O2
Molecular Weight: 350.46
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 151
Specific rotation [a]20/D: 118 deg (C=2, Pyridine)
Lambda max.: 315 nm (EtOH)

Formula: C₂₂H₂₆N₂O₂
Melting Pt: 151 °C
MDL Number: MFCD00211233
CAS Number: 42971-09-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50276f07890af5b438d9d90f67caae53bd0e0719&vid=50276f07890af5b438d9d90f67caae53bd0e0719&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR