Disodium-1-naphthol-3,6-disulfonate hydrate

Supplier: TCI America

Synonyms: Violet acid disodium salt, 4-Hydroxy-2,7-naphthalenedisulfonic acid disodium salt, Rudolf Guercke acid disodium salt, Disodium 4-Hydroxy-2,7-naphthalenedisulphonate, 1-Naphthol-3,6-disulfonic acid disodium salt, 1-Naphthol-3,6-disulphonic acid disodium salt, Disodium 4-Hydroxy-2,7-naphthalenedisulfonate, 4-Hydroxy-2,7-naphthalenedisulphonic acid disodium salt

V0037-25G
TCV0037-025GEA 28.5 CAD
TCV0037-025G
Disodium-1-naphthol-3,6-disulfonate hydrate
Disodium-1-naphthol-3,6-disulfonate hydrate

CAS Number: 20349-39-7
MDL Number: MFCD00003966
Molecular Formula: C10H8O7S2
Molecular Weight: 348.25
Form: Crystal
Color: White


Other Product Names / Synonyms: Disodium 4-Hydroxy-2,7-naphthalenedisulfonate, 4-Hydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt, 1-Naphthol-3,6-disulfonic Acid Disodium Salt, Rudolf Guercke Acid Disodium Salt, Violet Acid Disodium Salt

Formula: C₁₀H₈O₇S₂
MDL Number: MFCD00003966
CAS Number: 20349-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb0b4c9919b48ace99108aa7b2b79572c833db92&vid=eb0b4c9919b48ace99108aa7b2b79572c833db92&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR