2-Undecanone ≥98.0%

Supplier: TCI America

U0006-500ML U0006-25ML
TCU0006-500MLEA 170.64 CAD
TCU0006-500ML TCU0006-025ML
2-Undecanone ≥98.0%
2-Undecanone

CAS Number: 112-12-9
MDL Number: MFCD00009583
Molecular Formula: C11H22O
Molecular Weight: 170.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 120
Flash Point (°C): 106
Freezing point (°C): 12
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Methyl Nonyl Ketone

Formula: C₁₁H₂₂O
Boiling Pt: 120 °C
Melting Pt: ∼12 °C
Density: 0.83 (20 °C)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00009583
CAS Number: 112-12-9
EINECS: 203-937-5
UN: 3082
ADR: 9,III
Merck Index: 12,06178
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f85b3814b54ed7604df749a08a1e303a7445d528&vid=606f480ceb5c1c6de7005277c1894663cd815503&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR