2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T3638-1G T3638-200MG
TCT3638-1GEA 263.86 CAD
TCT3638-1G TCT3638-200MG
2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine ≥98.0% (by GC, titration analysis)
2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine
Formula: C₈H₅F₃N₄
MDL Number: MFCD26403657
CAS Number: 219508-27-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 157.0 to 161.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR