Teriflunomide ≥98.0% (by GC)

Supplier: TCI America
Warning

T3287-100MG T3287-1G
TCT3287-100MGEA 127.02 CAD
TCT3287-100MG TCT3287-1G
Teriflunomide ≥98.0% (by GC)
Teriflunomide
Formula: C₁₂H₉F₃N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00910058
CAS Number: 163451-81-8
Merck Index: 14,09165

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 227.0 to 231.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR