4-Methylphenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America

T2888-1G T2888-5G
TCT2888-1GEA 41.08 CAD
TCT2888-1G TCT2888-5G
4-Methylphenylboronic acid pinacol ester ≥98.0% (by GC)
4-Methylphenylboronic acid pinacol ester

CAS Number: 195062-57-8
MDL Number: MFCD03453662
Molecular Formula: C13H19BO2
Molecular Weight: 218.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 54


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: p-Tolylboronic Acid Pinacol Ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene

Formula: C₁₃H₁₉BO₂
Melting Pt: 54 °C
MDL Number: MFCD03453662
CAS Number: 195062-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9fec43fa59ae87be7507fd5dd7471e95387dbaf&vid=a9fec43fa59ae87be7507fd5dd7471e95387dbaf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR