1,1,3,3-Tetramethoxypropane ≥98.0% (by GC) for biochemical research

Supplier: TCI America

T2833-5G
TCT2833-5GEA 95.91 CAD
TCT2833-5G
1,1,3,3-Tetramethoxypropane ≥98.0% (by GC) for biochemical research
1,1,3,3-Tetramethoxypropane

[for Biochemical Research]
CAS Number: 102-52-3
MDL Number: MFCD00008488
Molecular Formula: C7H16O4
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 78
Flash Point (°C): 63
Specific Gravity (20/20): 1.00
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipid Peroxide Measurement Reagents
Other Product Names / Synonyms: Malonaldehyde Bis(dimethyl Acetal)

Formula: C₇H₁₆O₄
Boiling Pt: 78 °C
Density: 1.00 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00008488
CAS Number: 102-52-3
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03783a74aa701dfcf77e30de9d380cdb18539cfd&vid=4ec0640d0a7a27d1d37ddbc9858adbf6759f3bce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR