2',3',5'-Tri-O-acetyl-6-chloro-2-iodopurine riboside ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

T2691-1G T2691-5G
TCT2691-1GEA 95.91 CAD
TCT2691-1G TCT2691-5G
2',3',5'-Tri-O-acetyl-6-chloro-2-iodopurine riboside ≥97.0% (by HPLC, total nitrogen)
2',3',5'-Tri-O-acetyl-6-chloro-2-iodopurine riboside

CAS Number: 5987-76-8
MDL Number: MFCD03701153
Molecular Formula: C16H16ClIN4O7
Molecular Weight: 538.68
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 184


Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 6-Chloro-2-iodo-9-(2,3,5-tri-O-acetyl-beta-D-ribofuranosyl)purine

Formula: C₁₆H₁₆ClIN₄O₇
Melting Pt: 184 °C
MDL Number: MFCD03701153
CAS Number: 5987-76-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ddf9e36674c999bd5dc88073685b42a5dd9ade2&vid=0ddf9e36674c999bd5dc88073685b42a5dd9ade2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR