1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose ≥98.0%

Supplier: TCI America

T2607-25G T2607-5G
TCT2607-25GEA 353.76 CAD
TCT2607-25G TCT2607-5G
1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose ≥98.0%
1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose

CAS Number: 62211-93-2
MDL Number: MFCD08458459
Molecular Formula: C11H16O7
Molecular Weight: 260.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 67
Specific rotation [a]20/D: -21 deg (C=2, EtOH)

Formula: C₁₁H₁₆O₇
Melting Pt: 67 °C
MDL Number: MFCD08458459
CAS Number: 62211-93-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ada372d40ab49e513bb0f1dcc20514ee96d78615&vid=ada372d40ab49e513bb0f1dcc20514ee96d78615&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR