1-Iodo-4,4,4-trifluorobutane ≥98.0%

Supplier: TCI America

T2529-5G T2529-25G
TCT2529-5GEA 136.67 CAD
TCT2529-5G TCT2529-25G
1-Iodo-4,4,4-trifluorobutane ≥98.0%
1-Iodo-4,4,4-trifluorobutane

CAS Number: 461-17-6
MDL Number: MFCD00041543
Molecular Formula: C4H6F3I
Molecular Weight: 237.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 128
Flash Point (°C): 100
Specific Gravity (20/20): 1.86

Formula: C₄H₆F₃I
Boiling Pt: 128 °C
Density: 1.86 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00041543
CAS Number: 461-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45b62e9fc639426a6b343620d390348182592e0d&vid=45b62e9fc639426a6b343620d390348182592e0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR