Tetrakis(triphenylphosphine)platinum(0)

Supplier: TCI America

Synonyms: Tetrakis(triphenylphosphine)platinum

T2485-1G
TCT2485-1GEA 175.38 CAD
TCT2485-1G
Tetrakis(triphenylphosphine)platinum(0)
Tetrakis(triphenylphosphine)platinum (0)

CAS Number: 14221-02-4
MDL Number: MFCD00010014
Molecular Formula: C72H60P4Pt
Molecular Weight: 1244.25
Form: Crystal
Color: Slightly Pale Yellow
Storage Temperature: 0-10°C

Formula: C₇₂H₆₀P₄Pt
MW: 1244.24
MDL Number: MFCD00010014
CAS Number: 14221-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ccc63460e86a57c2edd041ba83ee95f7c0507cf&vid=4ccc63460e86a57c2edd041ba83ee95f7c0507cf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR