Trisodium-N-(1-carboxylatoethyl)iminodiacetate hydrate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

T2202-25G T2202-500G
TCT2202-25GEA 45.01 CAD
TCT2202-25G TCT2202-500G
Trisodium-N-(1-carboxylatoethyl)iminodiacetate hydrate ≥95.0% (by titrimetric analysis)
Trisodium-N-(1-carboxylatoethyl)iminodiacetate hydrate

CAS Number: 164462-16-2
Molecular Formula: C7H11NO6
Molecular Weight: 271.11
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: N,N-Bis(carboxylatomethyl)alanine Trisodium Salt, N-(1-Carboxylatoethyl)iminodiacetic Acid Trisodium Salt, Trisodium 2-Methylnitrilotriacetate, Methyl Glycine Diacetic Acid Trisodium Salt, MGDA

Formula: C₇H₁₁NO₆
CAS Number: 164462-16-2

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR