1-(p-Toluenesulfonyl)imidazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1-(P-Toluenesulfonyl)imidazole, 1-(P-Toluenesulphonyl)imidazole, 1-Tosylimidazole

T1985-5G T1985-25G
TCT1985-5GEA 59.49 CAD
TCT1985-5G TCT1985-25G
1-(p-Toluenesulfonyl)imidazole ≥98.0% (by HPLC, total nitrogen)
1-(p-Toluenesulphonyl)imidazole

CAS Number: 2232-08-8
MDL Number: MFCD00005285
Molecular Formula: C10H10N2O2S
Molecular Weight: 222.26
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 77


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Nucleic Acid Synthesis
Other Product Names / Synonyms: 1-Tosylimidazole

Formula: C₁₀H₁₀N₂O₂S
MW: 222.26 g/mol
Melting Pt: 77 °C
MDL Number: MFCD00005285
CAS Number: 2232-08-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19d69d9e3ddc778219e4bb8b2d44a63833417b41&vid=19d69d9e3ddc778219e4bb8b2d44a63833417b41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR