Tris(2-methoxyphenyl)bismuthine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

T1838-1G
TCT1838-1GEA 387.1 CAD
TCT1838-1G
Tris(2-methoxyphenyl)bismuthine ≥97.0% (by titrimetric analysis)
Tris(2-methoxyphenyl)bismuthine

CAS Number: 83724-41-8
MDL Number: MFCD00092705
Molecular Formula: C21H21BiO3
Molecular Weight: 530.38
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 163

Formula: C₂₁H₂₁BiO₃
Melting Pt: 163 °C
MDL Number: MFCD00092705
CAS Number: 83724-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e50c899b64adf9b6812d41b4f045a538ff9de1b8&vid=e50c899b64adf9b6812d41b4f045a538ff9de1b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR