Trinonylamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1826-5ML
TCT1826-5MLEA 418.23 CAD
TCT1826-5ML
Trinonylamine ≥98.0% (by titrimetric analysis)
Trinonylamine

CAS Number: 2044-22-6
MDL Number: MFCD00053817
Molecular Formula: C27H57N
Molecular Weight: 395.76
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 270
Specific Gravity (20/20): 0.81

Formula: C₂₇H₅₇N
Boiling Pt: 270 °C
Density: 0.81 (20 °C)
MDL Number: MFCD00053817
CAS Number: 2044-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6578492a6872f5c52ccff094c8dffeff8a34fb71&vid=6578492a6872f5c52ccff094c8dffeff8a34fb71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR