5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 5,10,15,20-Tetrakis(perfluorophenyl)porphyrin, 5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine, 5,10,15,20-Tetrakis(pentafluorophenyl)-21h,23h-porphine

T1730-100MG
TCT1730-100MGEA 259.12 CAD
TCT1730-100MG
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin ≥95.0% (by HPLC)
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin

CAS Number: 25440-14-6
MDL Number: MFCD00010032
Molecular Formula: C44H10F20N4
Molecular Weight: 974.56
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: Purple


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 5,10,15,20-Tetrakis(perfluorophenyl)porphyrin, 5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine

Formula: C₄₄H₁₀F₂₀N₄
MW: 974.56 g/mol
MDL Number: MFCD00010032
CAS Number: 25440-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ef30bfebedf1662cd8dd9877afee513ac155bac&vid=0ef30bfebedf1662cd8dd9877afee513ac155bac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR