Scandium (III) trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1663-1G T1663-5G
TCT1663-1GEA 90.85 CAD
TCT1663-1G TCT1663-5G
Scandium (III) trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Scandium (III) trifluoromethanesulphonate

CAS Number: 144026-79-9
MDL Number: MFCD00192433
Molecular Formula: C3F9O9S3Sc
Molecular Weight: 492.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Lewis Acids (Multi-component Reaction), Stable Lewis Acids in Aqueous Media
Other Product Names / Synonyms: Scandium(III) Triflate, Trifluoromethanesulfonic Acid Scandium(III) Salt

Formula: C₃F₉O₉S₃Sc
MDL Number: MFCD00192433
CAS Number: 144026-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c321a0d538d42c5ec08e6f73ad98aeeae0d81647&vid=c321a0d538d42c5ec08e6f73ad98aeeae0d81647&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR