(α,α,α-Trifluoro-3-tolyl)acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T1662-5G T1662-25G
TCT1662-005GEA 61.62 CAD
TCT1662-005G TCT1662-025G
(α,α,α-Trifluoro-3-tolyl)acetic acid ≥98.0% (by titrimetric analysis)
(α,α,α-Trifluoro-3-tolyl)acetic acid

CAS Number: 351-35-9
MDL Number: MFCD00004339
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 77


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: (alpha,alpha,alpha-Trifluoro-m-tolyl)acetic Acid

Formula: (C₆H₄(CF₃))CH₂COOH
Melting Pt: 77 °C
MDL Number: MFCD00004339
CAS Number: 351-35-9
EINECS: 206-511-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f99eab6b9545e3842b4fdd54d609a3fd85267c7&vid=4f99eab6b9545e3842b4fdd54d609a3fd85267c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR