3-Thiophenemethanol ≥96.0%

Supplier: TCI America

T1660-25G T1660-5G
TCT1660-25GEA 504.02 CAD
TCT1660-25G TCT1660-005G
3-Thiophenemethanol ≥96.0%
3-Thiophenemethanol

CAS Number: 71637-34-8
MDL Number: MFCD00014534
Molecular Formula: C5H6OS
Molecular Weight: 114.16
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 73
Flash Point (°C): 101
Specific Gravity (20/20): 1.22


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Hydroxymethylthiophene

Formula: C₅H₆OS
Boiling Pt: 73 °C
Density: 1.22 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00014534
CAS Number: 71637-34-8
EINECS: 275-741-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=177639f51230135415552cd80b62a0d5145e21b9&vid=177639f51230135415552cd80b62a0d5145e21b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR