TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

T1656-5G T1656-1G T1656-25G
TCT1656-5GEA 464.16 CAD
TCT1656-5G TCT1656-001G TCT1656-25G
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥98.0% (by titrimetric analysis)
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)

CAS Number: 51805-45-9
MDL Number: MFCD00145469
Molecular Formula: C9H15O6P
Molecular Weight: 286.64
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 177

Formula: C₉H₁₆ClO₆P
Melting Pt: 177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145469
CAS Number: 51805-45-9
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd5ee29f8f3e3f653187de712be2099096c31ce5&vid=cd5ee29f8f3e3f653187de712be2099096c31ce5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR