Tetrakis[N-phthaloyl-(S)-phenylalaninato]dirhodium ethyl acetate adduct

Supplier: TCI America

Synonyms: Rh2(S-PTPA)4 EtOAc, Dirhodium Tetrakis[N-phthaloyl-(S)-phenylalaninate] Ethyl Acetate Adduct

T1551-100MG
TCT1551-100MGEA 164.32 CAD
TCT1551-100MG
Tetrakis[N-phthaloyl-(S)-phenylalaninato]dirhodium ethyl acetate adduct
Tetrakis[N-phthaloyl-(S)-phenylalaninato]dirhodium ethyl acetate

CAS Number: 131219-55-1
MDL Number: MFCD00191695
Molecular Formula: C68H48N4O16Rh2
Molecular Weight: 1471.06
Form: Crystal
Color: Yellow Green


Other Product Names / Synonyms: Dirhodium Tetrakis[N-phthaloyl-(S)-phenylalaninate] Ethyl Acetate Adduct, Rh2(S-PTPA)4 EtOAc

Formula: C₆₈H₄₈N₄O₁₆Rh₂
MW: 1471.06 g/mol
MDL Number: MFCD00191695
CAS Number: 131219-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8226db4c6d948b70e5c8136a956d92afb3830b3&vid=a000faad5a6dea46af10b3cce5194f04e720eee3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR