2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0%

Supplier: TCI America

Synonyms: 1-(Anthracen-9-yl)-2,2,2-trifluoroethanol, alpha-(Trifluoromethyl)-9-anthracenemethanol, 1-(9-Anthryl)-2,2,2-trifluoroethanol

T1522-1G
TCT1522-001GEA 342.23 CAD
TCT1522-001G
2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0%
2,2,2-Trifluoro-1-(9-anthryl)ethanol

CAS Number: 65487-67-4
MDL Number: MFCD00062967
Molecular Formula: C16H11F3O
Molecular Weight: 276.26
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 142


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(9-Anthryl)-2,2,2-trifluoroethanol, alpha-(Trifluoromethyl)-9-anthracenemethanol

Formula: C₁₆H₁₁F₃O
MW: 276.26 g/mol
Melting Pt: 142 °C
MDL Number: MFCD00062967
CAS Number: 65487-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b26e2632274ac7b679e08a4191588315f6d0e0fe&vid=b26e2632274ac7b679e08a4191588315f6d0e0fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR